accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBrBrBr

CAS RN 75-80-9

IUPAC Name: 2,2,2-tribromoethan-1-ol
Synonyms: avertin
Molecular Weight (g/mol): 282.76
Molecular Formula: C2H3Br3O
InChi Key: YFDSDPIBEUFTMI-UHFFFAOYSA-N
SMILES: OCC(Br)(Br)Br
Molecular Weight (g/mol) 
  • (6)
  • (9)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (6)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (7)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (4)
  • (2)
  • (3)
  • (6)
  • (6)
  • (9)
  • (3)
  • (2)
  • (4)
  • (6)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)