Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Tribromoethanol, 99%

Catalog No. AAA1870614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1870614 25 g
AAA1870606 5 g
AAA1870622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1870614 Supplier Thermo Scientific Chemicals Supplier No. A1870614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 75-80-9 | C2H3Br3O | 282.757 g/mol

2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

Solubility
Soluble in alcohol, ether, benzene and warmpetroleum ether. Partially soluble in water.

Notes
Light Sensitive. Store in cool and well ventilated area. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-80-9
Molecular Formula C2H3Br3O
Molecular Weight (g/mol) 282.757
MDL Number MFCD00004671
InChI Key YFDSDPIBEUFTMI-UHFFFAOYSA-N
Synonym tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
PubChem CID 6400
IUPAC Name 2,2,2-tribromoethanol
SMILES C(C(Br)(Br)Br)O

Specifications

Melting Point 77°C to 80°C
Boiling Point 92°C to 93°C (10 mmHg)
Quantity 25 g
Beilstein 1733249
Sensitivity Light sensitive
Solubility Information Soluble in alcohol,ether,benzene and warmpetroleum ether. Partially soluble in water.
Formula Weight 282.76
Percent Purity 99%
Chemical Name or Material 2,2,2-Tribromoethanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-903-1
RTECSNumber KM3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.