accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COHO

CAS RN 75-91-2

IUPAC Name: 2-methylpropane-2-peroxol
Synonyms: tert-butyl hydroperoxide t-butylhydroperoxide tert-butylhydroperoxide t-butyl hydroperoxide hydroperoxyde de butyle tertiaire dimethylethyl hydroperoxide TBHP tertiary-butyl hydroperoxide
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: CIHOLLKRGTVIJN-UHFFFAOYSA-N
SMILES: CC(C)(C)OO
Molecular Weight (g/mol) 
  • (15)
  • (4)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
  • (1)
  • (3)
  • (5)
Percent Purity 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (8)
  • (4)
  • (1)
  • (6)
  • (15)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (5)
  • (8)
  • (5)
  • (2)
  • (4)