Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Hydroperoxide, 70% Solution in Water, Spectrum™ Chemical
SDP

Catalog No. 18607310 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-310 200 L Amber Glass Bottle
18-607-307 25 mL Amber Glass Bottle
18-607-308 100 mL Amber Glass Bottle
18-607-309 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-310 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B2499200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

B2499, 75-91-2, C4H10O2

Spectrum™ Chemical tert-Butyl Hydroperoxide, 70 Percent Solution in Water , with a systematic name of tert-Butyl hydroperoxide, is used in a variety of oxidation processes. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 75-91-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
IUPAC Name 2-methylpropane-2-peroxol
SMILES CC(C)(C)OO

Specifications

CAS Max % 1
Boiling Point 40°C (104°F)
Packaging Amber Glass Bottle
Quantity 200 L
UN Number UN3109
Grade Reagent
Physical Form Liquid
Chemical Name or Material tert-Butyl Hydroperoxide, 70 Percent Solution in Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.