accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOCH3OH3COCH3

CAS RN 7506-68-5

IUPAC Name: 3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-ol
Synonyms: 3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-ol
Molecular Weight (g/mol): 236.26
Molecular Formula: C10H20O6
InChi Key: AQWPITGEZPPXTJ-UHFFFAOYNA-N
SMILES: COCC1OC(O)C(OC)C(OC)C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results