Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetramethyl-D-glucose, MP Biomedicals

Catalog No. ICN222531 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222531 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222531 Supplier MP Biomedicals, Inc Supplier No. 0522253180

Chemical Identifiers

CAS 7506-68-5
Molecular Formula C10H20O6
Molecular Weight (g/mol) 236.264
InChI Key AQWPITGEZPPXTJ-YZUYTKQYSA-N
Synonym 2,3,4,6-tetra-o-methyl-d-glucopyranose, 2,3,4,6-tetra-o-methylhexopyranose #, d-glucopyranose, 2,3,4,6-tetramethyl, glucopyranose, 2,3,4,6-tetra-o-methyl-, d, 2,3,4,6-tetra-o-methylglucopyranose, 2-o,3-o,4-o,6-o-tetramethyl-d-glucopyranose, d-glucopyranose,2,3,4,6-tetra-o-methyl, 3r,4s,5r,6r-3,4,5-trimethoxy-6-methoxymethyl oxan-2-ol
PubChem CID 133126755
IUPAC Name (3S,4R,5S,6S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-ol
SMILES COCC1C(C(C(C(O1)O)OC)OC)OC

Specifications

Quantity 100 mg
Formula Weight 236
Chemical Name or Material 2,3,4,6-Tetramethyl-D-Glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.