accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OH3CH3CCH3

CAS RN 7507-48-4

IUPAC Name: 4-(acetyloxy)-2-tert-butylphenyl acetate
Synonyms: 4-(acetyloxy)-2-tert-butylphenyl acetate
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: SJRALGDWZXRRKL-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(OC(C)=O)C(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results