accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7521-80-4

IUPAC Name: butyltrichlorosilane
Synonyms: butyl trichloro silane
Molecular Weight (g/mol): 191.55
Molecular Formula: C4H9Cl3Si
InChi Key: FQEKAFQSVPLXON-UHFFFAOYSA-N
SMILES: CCCC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)