Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyltrichlorosilane 98.0+%, TCI America™
SDP

Catalog No. B039325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B039325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B039325G Supplier TCI America Supplier No. B039325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7521-80-4
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00013605
InChI Key FQEKAFQSVPLXON-UHFFFAOYSA-N
Synonym silane, butyltrichloro, n-butyltrichlorosilane, trichlorobutylsilane, unii-zq9o7lyo00, zq9o7lyo00, butylsilicon trichloride, n-butyl trichlorosilane, butyl trichlorosilane, hsdb 283, butyl-trichlorosilane
PubChem CID 24142
IUPAC Name butyl(trichloro)silane
SMILES CCCC[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
UN Number 1747
Formula Weight 191.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyltrichlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.