Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyltrichlorosilane, 99%

Catalog No. AC435600250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435600250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435600250 Supplier Thermo Scientific Chemicals Supplier No. 435600250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7521-80-4 | C4H9Cl3Si | 191.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7521-80-4
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.56
InChI Key FQEKAFQSVPLXON-UHFFFAOYSA-N
Synonym silane, butyltrichloro, n-butyltrichlorosilane, trichlorobutylsilane, unii-zq9o7lyo00, zq9o7lyo00, butylsilicon trichloride, n-butyl trichlorosilane, butyl trichlorosilane, hsdb 283, butyl-trichlorosilane
PubChem CID 24142
IUPAC Name butyl(trichloro)silane
SMILES CCCC[Si](Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.1610g/mL
Boiling Point 142°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380 (589
Linear Formula CH3(CH2)3SiCl3
Quantity 25 mL
Specific Gravity 1.161
Solubility Information Solubility in water: reacts
Formula Weight 191.56
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyltrichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Store in a dry place.
Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rin
EINECSNumber 231-381-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.