accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOCH3ClOHN(R)N(R)H3C

CAS RN 75272-39-8

IUPAC Name: N-[(2R,3R)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide
Synonyms: (+)-nemonapride (2R,3R)-nemonapride
Molecular Weight (g/mol): 387.91
Molecular Formula: C21H26ClN3O2
InChi Key: KRVOJOCLBAAKSJ-RDTXWAMCSA-N
SMILES: CNC1=CC(OC)=C(C=C1Cl)C(=O)N[C@@H]1CCN(CC2=CC=CC=C2)[C@@H]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)