accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75308-12-2

IUPAC Name: 2-dodecylfuran
Synonyms: 2-dodecylfuran
Molecular Weight (g/mol): 236.4
Molecular Formula: C16H28O
InChi Key: YCHOIEWPPPNUFM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results