accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHSnHClCH2H2C

CAS RN 7532-85-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 245.72
Molecular Formula: C4H8Cl2Sn
InChi Key: OFJXSWMLVIFLKC-UHFFFAOYSA-N
SMILES: Cl.Cl.C=C[SnH2]C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results