accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 75359-31-8

IUPAC Name: 1,2-ditridecyl benzene-1,2-dicarboxylate
Synonyms: ditridecyl phthalate
Molecular Weight (g/mol): 530.83
Molecular Formula: C34H58O4
InChi Key: YCZJVRCZIPDYHH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results