CAS RN 754954-71-7
IUPAC Name:
5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms:
5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol):
795.7
Molecular Formula:
C31H45N3O21
InChi Key:
OQZSJFGKSKBLDR-UHFFFAOYNA-N
SMILES:
CC(=O)NC1C(O)CC(OCC2OC(OC3=CC=C(C=C3)[N+]([O-])=O)C(NC(C)=O)C(OC3OC(CO)C(O)C(O)C3O)C2O)(OC1C(O)C(O)CO)C(O)=O
1
–
1
of
1
results
1
Gal beta(1-3)[Neu5Ac alpha(2-6)]GlcNAc-beta-pNP 92.0+%, TCI America™
CAS: 754954-71-7 Molecular Formula: C31H45N3O21 Molecular Weight (g/mol): 795.70 MDL Number: MFCD15072169 InChI Key: OQZSJFGKSKBLDR-UHFFFAOYNA-N PubChem CID: 133554203 IUPAC Name: 5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid SMILES: CC(=O)NC1C(O)CC(OCC2OC(OC3=CC=C(C=C3)[N+]([O-])=O)C(NC(C)=O)C(OC3OC(CO)C(O)C(O)C3O)C2O)(OC1C(O)C(O)CO)C(O)=O