Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gal beta(1-3)[Neu5Ac alpha(2-6)]GlcNAc-beta-pNP 92.0+%, TCI America™
SDP

Catalog No. G03472MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03472MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03472MG Supplier TCI America Supplier No. G03472MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 754954-71-7
Molecular Formula C31H45N3O21
Molecular Weight (g/mol) 795.70
MDL Number MFCD15072169
InChI Key OQZSJFGKSKBLDR-UHFFFAOYNA-N
PubChem CID 133554203
IUPAC Name 5-acetamido-2-{[5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]methoxy}-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
SMILES CC(=O)NC1C(O)CC(OCC2OC(OC3=CC=C(C=C3)[N+]([O-])=O)C(NC(C)=O)C(OC3OC(CO)C(O)C(O)C3O)C2O)(OC1C(O)C(O)CO)C(O)=O

Specifications

Quantity 2 mg
Formula Weight 795.70
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-3)[Neu5Ac alpha(2-6)]GlcNAc-beta-pNP
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.