accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75507-26-5

IUPAC Name: [(2S)-1,4,7,10-tetraoxacyclododecan-2-yl]methanol
Synonyms: (2S)-1,4,7,10-tetraoxacyclododecan-2-ylmethanol
Molecular Weight (g/mol): 206.24
Molecular Formula: C9H18O5
InChi Key: NJIPEIQHUNDGPY-VIFPVBQESA-N
SMILES: OC[C@H]1COCCOCCOCCO1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)