accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75618-25-6

IUPAC Name: butyl trifluoromethanesulfonate
Synonyms: butyl trifluoromethanesulfonate
Molecular Weight (g/mol): 206.18
Molecular Formula: C5H9F3O3S
InChi Key: SEXLAQXMAFCJCO-UHFFFAOYSA-N
SMILES: CCCCOS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results