CAS RN 75621-03-3
Thermo Scientific™ VitroEase™ Buffer Screening Kit and Detergents
The Thermo Scientific VitroEase Buffer Screening Kit provides pre-formulated buffers and detergents and a comprehensive screening strategy to identify optimal protein conditions for single particle cryo-EM sample analysis.
CHAPS (White Crystalline Powder), Fisher BioReagents™
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPS, MP Biomedicals™
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.88 InChI Key: UMCMPZBLKLEWAF-BCTGSCMUSA-N Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate,CHAPS hydrate
Chaps, 99.8%, MP Biomedicals™
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)-dimethyl- ammonio]-1-propane sulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPS, 98+%
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 MDL Number: MFCD00012116 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
MilliporeSigma™ CHAPS, OmniPur™, Calbiochem™,
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)-dimethylammino]-propane- sulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPSO , MP Biomedicals
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)-dimethyl- ammonio]-2-hydroxy-1-propane sulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPS, MP Biomedicals
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 MDL Number: MFCD00012116 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl) dimethylammonio]-1-propane sulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPS, Electrophoresis Grade
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 MDL Number: MFCD00012116 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
MilliporeSigma™ CHAPS, Molecular biology grade, Calbiochem™,
CAS: 75621-03-3
Thermo Scientific Chemicals CHAPS
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 MDL Number: MFCD00012116 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPS, White crystalline powder, Hoefer™
CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Molecular Weight (g/mol): 614.883 InChI Key: UMCMPZBLKLEWAF-GBSSQDIOSA-N Synonym: 3-[(3-Cholamidopropyl)-Dimethylammonio]-1-Propane Sulfonate PubChem CID: 134129639 IUPAC Name: 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate SMILES: CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C