Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chaps, 99.8%, MP Biomedicals™

Catalog No. ICN19031980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19031980 100 g
ICN19031901 1 g
ICN19031925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19031980 Supplier MP Biomedicals, Inc Supplier No. 0219031980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)-dimethyl- ammonio]-1-propane sulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Physical Form Crystalline Powder
pH 4.0 to 7.0
Quantity 100 g
Assay Percent Range 0.998
Formula Weight 614.9
Percent Purity ≥98%
Grade Ungraded
Chemical Name or Material Chaps
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.