accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C(?)

CAS RN 75640-87-8

IUPAC Name: 2,2'-dimethoxy-1,1'-binaphthalene
Synonyms: 2,2'-dimethoxy-1,1'-binaphthalene
Molecular Weight (g/mol): 314.38
Molecular Formula: C22H18O2
InChi Key: BJAADAKPADTRCH-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results