Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2,2'-Dimethoxy-1,1'-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. D28211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28211G Supplier TCI America Supplier No. D28211G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75640-87-8
Molecular Formula C22H18O2
Molecular Weight (g/mol) 314.384
MDL Number MFCD00091146
InChI Key BJAADAKPADTRCH-UHFFFAOYSA-N
Synonym (S)-1,1′C-Bi-2-naphthol Dimethyl Ether
PubChem CID 235616
IUPAC Name 2-methoxy-1-(2-methoxynaphthalen-1-yl)naphthalene
SMILES COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OC

Specifications

Melting Point 230°C
Color White
Quantity 1 g
Formula Weight 314.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2,2′-Dimethoxy-1,1′-binaphthyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.