accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNOOOOOOH

CAS RN 756525-91-4

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}-3,6,9,12-tetraoxapentadecan-15-oic acid
Synonyms: 1-[(tert-butoxycarbonyl)amino]-3,6,9,12-tetraoxapentadecan-15-oic acid
Molecular Weight (g/mol): 365.42
Molecular Formula: C16H31NO8
InChi Key: YEIYIPDFZMLJQH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCOCCOCCOCCOCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)