Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Boc-amino)-PEG4-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B5665250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5665250MG 250 mg
B56651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5665250MG Supplier TCI America Supplier No. B5665250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 756525-91-4
Molecular Formula C16H31NO8
Molecular Weight (g/mol) 365.423
MDL Number MFCD07781254
InChI Key YEIYIPDFZMLJQH-UHFFFAOYSA-N
Synonym 15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic Acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic Acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic Acid, Carboxy-PEG4-(Boc-amine)
PubChem CID 2756001
IUPAC Name 3-[2-[2-[2-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
SMILES CC(C)(C)OC(=O)NCCOCCOCCOCCOCCC(=O)O

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 365.42
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Liquid
Chemical Name or Material (Boc-amino)-PEG4-carboxylic Acid
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.