CAS RN 757949-98-7
IUPAC Name:
2-[2-(3-fluorophenyl)ethynyl]-4,6-dimethylpyridin-3-amine hydrochloride
Synonyms:
raseglurant hydrochloride
Molecular Weight (g/mol):
276.74
Molecular Formula:
C15H14ClFN2
InChi Key:
BDXXMGYKLQZXOU-UHFFFAOYSA-N
SMILES:
Cl.CC1=NC(C#CC2=CC(F)=CC=C2)=C(N)C(C)=C1
1
–
1
of
1
results
1
R&D Systems™ ADX 10059 hydrochloride
CAS: 757949-98-7 Molecular Formula: C15H14ClFN2