accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 76-01-7

IUPAC Name: 1,1,1,2,2-pentachloroethane
Synonyms: pentachloroethane
Molecular Weight (g/mol): 202.28
Molecular Formula: C2HCl5
InChi Key: BNIXVQGCZULYKV-UHFFFAOYSA-N
SMILES: ClC(Cl)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results