Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentachloroethane Solution, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USCH2301
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-CH-230-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-CH-230-1 Supplier Agilent Technologies Supplier No. CH2301
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 76-01-7 , 67-56-1
Molecular Formula C2HCl5
Molecular Weight (g/mol) 202.28
InChI Key BNIXVQGCZULYKV-UHFFFAOYSA-N
PubChem CID 6419
ChEBI CHEBI:76287
IUPAC Name 1,1,1,2,2-pentachloroethane
SMILES C(C(Cl)(Cl)Cl)(Cl)Cl

Specifications

CAS Max % 99.98, 0.01
Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Pentachloroethane, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.