CAS RN 76-06-2
IUPAC Name:
trichloro(nitro)methane
Synonyms:
chloropicrin trichloronitromethane (chloropicrine) chlorpikrin 1,1,1-trichloronitromethane
Molecular Weight (g/mol):
164.37
Molecular Formula:
CCl3NO2
InChi Key:
LFHISGNCFUNFFM-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C(Cl)(Cl)Cl
1
–
1
of
1
results
1
Chloropicrin, SPEX CertiPrep™
CAS: 76-06-2 Molecular Formula: CCl3NO2 Molecular Weight (g/mol): 164.366 InChI Key: LFHISGNCFUNFFM-UHFFFAOYSA-N PubChem CID: 6423 ChEBI: CHEBI:39285 IUPAC Name: trichloro(nitro)methane SMILES: C([N+](=O)[O-])(Cl)(Cl)Cl