accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNNCS(E/Z)

CAS RN 7612-98-8

IUPAC Name: 4-[2-(4-isothiocyanatophenyl)diazen-1-yl]-N,N-dimethylaniline
Synonyms: 4-[2-(4-isothiocyanatophenyl)diazen-1-yl]-N,N-dimethylaniline
Molecular Weight (g/mol): 282.37
Molecular Formula: C15H14N4S
InChi Key: OSWZKAVBSQAVFI-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)N=C=S
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)