Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)azobenzene 4'-Isothiocyanate 95.0+%, TCI America™
SDP

Catalog No. D1653100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1653100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1653100MG Supplier TCI America Supplier No. D1653100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7612-98-8
Molecular Formula C15H14N4S
Molecular Weight (g/mol) 282.37
MDL Number MFCD00004812
InChI Key OSWZKAVBSQAVFI-UHFFFAOYSA-N
Synonym dabitc, 4-n,n-dimethylamino azobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenylazo-n,n-dimethylaniline, 4-dimethylaminoazobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenyl azo-n,n-dimethylaniline, 4-dimethylamino azobenzene 4'-isothiocyanate, 4-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-e-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-n,n-dimethylaminoazobenzene-4'-isothiocyanate, 4-dimethylamino-4'-isothiocyanatoazobenzene
PubChem CID 82084
IUPAC Name 4-[2-(4-isothiocyanatophenyl)diazen-1-yl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 169°C
Quantity 100 mg
Formula Weight 282.37
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)azobenzene 4′-Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.