Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminoazobenzene-4'-isothiocyanate, 97%, Thermo Scientific™

Catalog No. AAA1258103
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1258103 1 g
AAA1258101 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1258103 Supplier Thermo Scientific Chemicals Supplier No. A1258103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7612-98-8
Molecular Formula C15H14N4S
Molecular Weight (g/mol) 282.37
MDL Number MFCD00004812
InChI Key OSWZKAVBSQAVFI-UHFFFAOYSA-N
Synonym dabitc, 4-n,n-dimethylamino azobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenylazo-n,n-dimethylaniline, 4-dimethylaminoazobenzene-4'-isothiocyanate, 4-4-isothiocyanatophenyl azo-n,n-dimethylaniline, 4-dimethylamino azobenzene 4'-isothiocyanate, 4-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-e-2-4-isothiocyanatophenyl diazen-1-yl-n,n-dimethylaniline, 4-n,n-dimethylaminoazobenzene-4'-isothiocyanate, 4-dimethylamino-4'-isothiocyanatoazobenzene
PubChem CID 82084
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 169°C to 171°C
Quantity 1 g
Beilstein 752568
Sensitivity Moisture sensitive
IUPAC Name 4-[2-(4-isothiocyanatophenyl)diazen-1-yl]-N,N-dimethylaniline
Formula Weight 282.37
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-isothiocyanate
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-521-3
RTECSNumber NX9125000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.