accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 761446-44-0

IUPAC Name: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 208.07
Molecular Formula: C10H17BN2O2
InChi Key: UCNGGGYMLHAMJG-UHFFFAOYSA-N
SMILES: CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)