CAS RN 76153-06-5
CAS RN 76153-06-5
IUPAC Name:
5-bromo-3-methylbenzene-1,2-diamine
Synonyms:
5-bromo-3-methylbenzene-1,2-diamine
Molecular Weight (g/mol):
201.07
Molecular Formula:
C7H9BrN2
InChi Key:
UOFSLKHZOPVGHG-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=CC(N)=C1N
1
–
1
of
1
results
1
5-Bromo-3-methyl-benzene-1,2-diamine, 97%, Thermo Scientific™
CAS: 76153-06-5 Molecular Formula: C7H9BrN2 Molecular Weight (g/mol): 201.067 InChI Key: UOFSLKHZOPVGHG-UHFFFAOYSA-N Synonym: 5-bromo-3-methyl-benzene-1,2-diamine,5-bromo-3-methyl-1,2-benzenediamine,1,2-benzenediamine, 5-bromo-3-methyl,2,3-diamino-5-bromotoluene,5-bromo-2,3-diaminotoluene,bromomethylbenzenediamine,pubchem22667,ksc642k2n,5-bromo-2,3-diamino-toluene,5-bromo-2.3-diamino-toluene PubChem CID: 1516397 IUPAC Name: 5-bromo-3-methylbenzene-1,2-diamine SMILES: CC1=CC(=CC(=C1N)N)Br