accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNHNH2OHO

CAS RN 7622-29-9

IUPAC Name: hydrogen 2-amino-6-(methylamino)hexanoic acid chloride
Synonyms: hydrogen 2-amino-6-(methylamino)hexanoic acid chloride
Molecular Weight (g/mol): 196.68
Molecular Formula: C7H17ClN2O2
InChi Key: AQELUQTVJOFFBN-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNCCCCC(N)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)