accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76397-53-0

IUPAC Name: 1-(2-sulfanylidene-1,3-thiazolidin-3-yl)ethan-1-one
Synonyms: 1-(2-sulfanylidene-1,3-thiazolidin-3-yl)ethanone
Molecular Weight (g/mol): 161.24
Molecular Formula: C5H7NOS2
InChi Key: BMOBFBQWISCVKX-UHFFFAOYSA-N
SMILES: CC(=O)N1CCSC1=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results