accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONOONOO

CAS RN 7659-29-2

IUPAC Name: 3,5-dinitrobenzene-1,2-bis(olate)
Synonyms: 3,5-dinitrobenzene-1,2-bis(olate)
Molecular Weight (g/mol): 198.09
Molecular Formula: C6H2N2O6
InChi Key: VDCDWNDTNSWDFJ-UHFFFAOYSA-L
SMILES: [O-]C1=CC(=CC(=C1[O-])[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)