accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 76709-55-2

IUPAC Name: nonadeca-2,4-diynoic acid
Synonyms: nonadeca-2,4-diynoic acid
Molecular Weight (g/mol): 290.45
Molecular Formula: C19H30O2
InChi Key: LFKNZQNITZRUEE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC#CC#CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results