accessibility menu, dialog, popup

UserName

Viewing profile as user:
PFFFFFFFFFFPFFFFFFFFFF

CAS RN 76858-94-1

IUPAC Name: {2-[bis(2,3,4,5,6-pentafluorophenyl)phosphanyl]ethyl}bis(2,3,4,5,6-pentafluorophenyl)phosphane
Synonyms: {2-[bis(2,3,4,5,6-pentafluorophenyl)phosphanyl]ethyl}bis(2,3,4,5,6-pentafluorophenyl)phosphane
Molecular Weight (g/mol): 758.23
Molecular Formula: C26H4F20P2
InChi Key: IGLFIYOFKVGEBP-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(P(CCP(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)