Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dipentafluorophenylphosphino)ethane, 99%, Thermo Scientific™

Catalog No. AA3927606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3927606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3927606 Supplier Thermo Scientific Chemicals Supplier No. 3927606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76858-94-1
Molecular Formula C26H4F20P2
Molecular Weight (g/mol) 758.23
MDL Number MFCD00058841
InChI Key IGLFIYOFKVGEBP-UHFFFAOYSA-N
Synonym 1,2-bis dipentafluorophenylphosphino ethane, 1,2-bis bis pentafluorophenyl phosphino ethane, 1,2-bis bis perfluorophenyl phosphino ethane, 2-bis 2,3,4,5,6-pentafluorophenyl phosphanylethyl-bis 2,3,4,5,6-pentafluorophenyl phosphane, ethane-1,2-diylbis bis pentafluorophenyl phosphane, dfppe, pubchem6546, acmc-209p6f, ethylenebis bis pentafluorophenyl phosphine, bis dipentafluorophenylphosphine ethane
PubChem CID 2734566
IUPAC Name 2-bis(2,3,4,5,6-pentafluorophenyl)phosphanylethyl-bis(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(CCP(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 185°C to 190°C
Color White
Linear Formula (C6F5)2PCH2CH2P(C6F5)2
Quantity 5 g
UN Number UN3464
Solubility Information Insoluble in water.
Formula Weight 758.23
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material 1,2-Bis(dipentafluorophenylphosphino)ethane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.