Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis[bis(pentafluorophenyl)phosphino]ethane 97.0+%, TCI America™
SDP

Catalog No. B34281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34281G Supplier TCI America Supplier No. B34281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76858-94-1
Molecular Formula C26H4F20P2
Molecular Weight (g/mol) 758.23
MDL Number MFCD00058841
InChI Key IGLFIYOFKVGEBP-UHFFFAOYSA-N
Synonym 1,2-bis dipentafluorophenylphosphino ethane, 1,2-bis bis pentafluorophenyl phosphino ethane, 1,2-bis bis perfluorophenyl phosphino ethane, 2-bis 2,3,4,5,6-pentafluorophenyl phosphanylethyl-bis 2,3,4,5,6-pentafluorophenyl phosphane, ethane-1,2-diylbis bis pentafluorophenyl phosphane, dfppe, pubchem6546, acmc-209p6f, ethylenebis bis pentafluorophenyl phosphine, bis dipentafluorophenylphosphine ethane
PubChem CID 2734566
IUPAC Name {2-[bis(2,3,4,5,6-pentafluorophenyl)phosphanyl]ethyl}bis(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(CCP(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 195°C
Color White
Quantity 1 g
Formula Weight 758.23
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis[bis(pentafluorophenyl)phosphino]ethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.