accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNH2NBrOHOPOOHONa

CAS RN 76939-46-3

IUPAC Name: sodium 6-(6-amino-8-bromo-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Synonyms: sodium 6-(6-amino-8-bromopurin-9-yl)-2,7-dihydroxy-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Molecular Weight (g/mol): 431.09
Molecular Formula: C10H11BrN5NaO6P
InChi Key: DMRMZQATXPQOTP-UHFFFAOYNA-N
SMILES: [Na+].NC1=C2N=C(Br)N(C3OC4COP(O)(=O)OC4C3O)C2=NC=N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results