CAS RN 77-09-8
Phenolphthalein (Certified ACS), Fisher Chemical™
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N Synonym: 3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone,3, 3-Bis(p-hydroxyphenyl) Phthallide PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenolphthalein, ACS reagent
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenolphthalein, 2% soln. in 95% ethanol
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N Synonym: phenolphthalein,phthalimetten,euchessina,phthalin,espotabs,phenolax,purgophen,koprol,laxogen,trilax PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenolphthalein, ACS
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenolphthalein (0.1% in ca. 90% Ethanol) [for Titration], TCI America™
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N Synonym: phenolphthalein,phthalimetten,euchessina,phthalin,espotabs,phenolax,purgophen,koprol,laxogen,trilax PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenolphthalein, 98%
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N Synonym: phenolphthalein,phthalimetten,euchessina,phthalin,espotabs,phenolax,purgophen,koprol,laxogen,trilax PubChem CID: 4764 ChEBI: CHEBI:34914 SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Thermo Scientific Chemicals Phenolphthalein, 98.5%, pure, Indicator grade
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenolphthalein, 1% (w/v) Indicator Solution in 95% Ethanol, Spectrum™ Chemical
Phenolphthalein Solution, Sigma-Aldrich
Synthetic acid-base indicator that remains colorless in acidic or neutral solutions and turns pink in alkaline environments.
Thermo Scientific Chemicals Phenolphthalein, 0.5% w/v in alcohol
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenolphthalein, 1% (w/v) Indicator Solution in 70% Alcohol, Spectrum™ Chemical
Phenolphthalein StandAR, Macron Fine Chemicals™
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Phenolphthalein StandAR, Macron Fine Chemicals™
CAS: 77-09-8 Molecular Formula: C20H14O4 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00005913 InChI Key: KJFMBFZCATUALV-UHFFFAOYSA-N PubChem CID: 4764 ChEBI: CHEBI:34914 IUPAC Name: 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one SMILES: OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1