CAS RN 7704-34-9
Sulfur, 99.5%, sublimed
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur powder, precipitated, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur, 99.5+%, refined
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur in Crude Oil standard solution, 1000μg/g (0.10%), Specpure™
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur pieces, Puratronic™, 99.999% (metals basis)
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Precipitated Sulfur, 99.5-100.5%, Spectrum™ Chemical
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N IUPAC Name: sulfur SMILES: [S]
Sulfur pieces, Puratronic™, 99.9995% (metals basis)
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur, 99.999%, (trace metal basis)
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur powder, -325 mesh, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur powder, sublimed, -100 mesh, 99.5%
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N Synonym: polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
Sulfur Oil Standard in No. 2 Diesel Fuel, 500μg/g, SPEX CertiPrep™
CAS: 7704-34-9 Molecular Formula: S Molecular Weight (g/mol): 32.06 MDL Number: MFCD00085316 InChI Key: NINIDFKCEFEMDL-UHFFFAOYSA-N PubChem CID: 5362487 ChEBI: CHEBI:26833 IUPAC Name: sulfur SMILES: [S]
- 1
- 2