Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Precipitated Sulfur, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605786 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-786 50 kg Poly Bottle
18-605-783 500 g Poly Bottle
18-605-784 2.5 kg Poly Bottle
18-605-785 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-786 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S138450KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1384, 7704-34-9, S

Spectrum™ Chemical Precipitated Sulfur is an abundant, non-metal, multivalent chemical element. In cosmetics and personal care products Sulfur is used to enhance the appearance and feel of hair, by increasing hair body, suppleness, or sheen, or by improving the texture of hair that has been damaged physically or by chemical treatment and to enhance the appearance of dry or damaged skin by reducing flaking and restoring suppleness.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
IUPAC Name sulfur
SMILES [S]

Specifications

CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 50 kg
Percent Purity 99.5 to 100.5%
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material Precipitated Sulfur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.