accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7705-07-9

IUPAC Name: titanium(3+) trichloride
Synonyms: titanium(III) chloride
Molecular Weight (g/mol): 154.22
Molecular Formula: Cl3Ti
InChi Key: YONPGGFAJWQGJC-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Ti+3]
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)