accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSClO

CAS RN 7719-09-7

IUPAC Name: sulfurooyl dichloride
Synonyms: thionyl chloride socl2 sulfinyl chloride thionyl dichloride sulfurous oxychloride sulfur oxychloride sulfur chloride oxide sulfinyl dichloride sulfur dichloride oxide
Molecular Weight (g/mol): 118.96
Molecular Formula: Cl2OS
InChi Key: FYSNRJHAOHDILO-UHFFFAOYSA-N
SMILES: ClS(Cl)=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)