Learn More
Description
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Percent Purity | 97% |
| Grade | Purified |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Thionyl Chloride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
