Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionyl Chloride, Purified, 97%, Spectrum™ Chemical
SDP

Catalog No. 18607382 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-382 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-382 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TH1381LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TH138, 7719-09-7, SOCl2

Spectrum™ Chemical Thionyl Chloride, Purified is a moderately volatile colorless liquid with an unpleasant acrid odor. Thionyl chloride is commonly used as a chlorinating reagent in the preparation of organic synthetic intermediates used in the preparation of pharmaceuticals and agrichemical products. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS 7719-09-7
Molecular Formula Cl2OS
Molecular Weight (g/mol) 118.96
InChI Key FYSNRJHAOHDILO-UHFFFAOYSA-N
SMILES ClS(Cl)=O

Specifications

CAS Max % 1
Physical Form Liquid
Quantity 1 L
Percent Purity 97%
Grade Purified
Packaging Amber Glass Bottle
Chemical Name or Material Thionyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.