CAS RN 772-54-3
IUPAC Name:
benzyldiethylamine
Synonyms:
benzyldiethylamine
Molecular Weight (g/mol):
163.26
Molecular Formula:
C11H17N
InChi Key:
ZWRDBWDXRLPESY-UHFFFAOYSA-N
SMILES:
CCN(CC)CC1=CC=CC=C1
1
–
1
of
1
results
1
N,N-Diethylbenzylamine 98.0+%, TCI America™
CAS: 772-54-3 Molecular Formula: C11H17N Molecular Weight (g/mol): 163.26 MDL Number: MFCD00026731 InChI Key: ZWRDBWDXRLPESY-UHFFFAOYSA-N Synonym: N-Benzyldiethylamine PubChem CID: 61211 IUPAC Name: benzyldiethylamine SMILES: CCN(CC)CC1=CC=CC=C1