Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylbenzylamine 98.0+%, TCI America™
SDP

Catalog No. D110325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110325ML 25 mL
D1103500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D110325ML Supplier TCI America Supplier No. D110325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 772-54-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.26
MDL Number MFCD00026731
InChI Key ZWRDBWDXRLPESY-UHFFFAOYSA-N
Synonym N-Benzyldiethylamine
PubChem CID 61211
IUPAC Name benzyldiethylamine
SMILES CCN(CC)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 mL
UN Number 2735
Formula Weight 163.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylbenzylamine
EINECSNumber (3)-0350
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.